R-Phycoerythrin (RPE)
Supplier: Columbia Biosciences
Certificates
About this item
Many applications and instruments were developed specifically for R-phycoerythrin.
It is commonly used in immunoassays such as FACS, flow cytometry, multimer/tetramer applications. With new instrumetation available, R-PE is also well suited for immunohistochemistry (IHC) and Western blot applications.
Specifications
- Storage temperature:Store at 4 °C
- Application:Flow cytometry, Cell-based assays, microarrays and microplate applications