Order Entry
Germany
ContactUsLinkComponent
DEAE Sephadex™ A-25 Ion Exchange Chromatography Media, Cytiva
DEAE Sephadex™ A-25 Ion Exchange Chromatography Media, Cytiva
Supplier:  Cytiva
Certificates
undefined
DEAE Sephadex™ A-25 Ion Exchange Chromatography Media, Cytiva
Supplier:  Cytiva

Specifications

  • pH range:
    2 - 9
  • Matrix:
    Cross-linked dextran
  • Ligand:
    Diethylaminoethyl
  • Storage temperature:
    4 - 30 °C
  • Binding capacity:
    Thyroglobulin (Mr 669 000)- 1 mg/ml drained medium), HSA (Mr 68 000)- 30 mg/ml drained medium), α-lactalbumin (Mr 14 300)- 140 mg/ml drained medium)
  • Total ionic capacity:
    3-4 mmol/g
  • Description:
    DEAE Sephadex™ A-25

Specifications