ContactUsLinkComponent
Serological Pipettes Holder
Serological Pipettes Holder
Catalog # KART0111500
Supplier:  KARTELL
undefined
Serological Pipettes Holder
Catalog # KART0111500
Supplier:  KARTELL
Specifications
  • Description:
    Holder for serological pipettes
  • W×D×H:
    300×100×420 mm
  • Pk:
    1
Specifications
About this item
Bench pipette holder, made of particularly clear polymethylmethacrylate (PMMA), has high level finishes and its content can be seen from any angle.
  • Complete with four tilted support shelves and white top
The four separate compartments are ideal for housing plastic or glass serologic pipettes of various capacities (from 1 to 50 ml). Do not shine ultraviolet light on it.