Order Entry
Germany
ContactUsLinkComponent
VWR® Vial Racks
VWR® Vial Racks
Catalog # 548-1388A
Supplier:  VWR Collection
VWR® Vial Racks
Catalog # 548-1388A
Supplier:  VWR Collection

Specifications

  • No. of Holes:
    50
  • Description:
    PP vial rack for 1,5 ml vials
  • W×D×H:
    200×105×17 mm
  • Pk:
    5

Specifications

About this item