Autoclavable Cleanroom Goggles, Isoclave IV
111-2618
- Colour:Clear
- Description:Isoclave IV autoclavable cleanroom goggles
- Lenses:Polycarbonate
- Pk:50
GMP Annex 1 compatible autoclavable goggle with indirect venting.
- Autoclavable cleanroom goggle
- ISO Class 5, GMP Annex 1 compatible
- Anti-fog coated lens (up to 35 cycles)
- Suitable for use over glasses
- Non-linting strap with adjustible side clips
- Indirect venting to prevent particle egress
The Isoclave IV google is made with a super soft thermoplastic housing to ensure comfort, and a wide view anti-fog coated lens. Suitable for autoclaving at 121 degrees Celsius for 30 minutes, the Isoclave offers up to 35 cycles on the lens and 40 on straps and housing, under lab conditions.
: PPE Cat II EN 166:2001. ISO 9001:2015.
: Each piece is sealed in HDPE bag. 10 sealed bags in 1 inner box, 5 inner boxes per case (50 pieces).