ContactUsLinkComponent
Quanti-Cult Plus™ Clostridium sporogenes ATCC™ 11437™
Quanti-Cult Plus™ Clostridium sporogenes ATCC™ 11437™
Catalog # OXOIR4711703
CAS Number:  
undefined
Quanti-Cult Plus™ Clostridium sporogenes ATCC™ 11437™
Catalog # OXOIR4711703
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Clostridium sporogenes ATCC™ 11437™
Specifications
About this item
Quanti-Cult Plus™ Clostridium sporogenes ATCC™ 11437™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0,1 ml inoculum
  • Ready for rehydration
  • For use in quality control procedures
Packaging: Includes 10 preserved organism vials and 10 rehydration vials.