Order Entry
Germany
ContactUsLinkComponent
Hydrochloric acid 37% NF
Hydrochloric acid 37% NF
Catalog # SPCMHY104-500MLGL
Supplier:  Spectrum Chemical
CAS Number:  
Hydrochloric acid 37% NF
Catalog # SPCMHY104-500MLGL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Assay (w/w):
    36.5 - 38.0%
  • Bromide or Iodide:
    To Pass Test
  • Free Bromine or Chlorine:
    To Pass Test

Specification Test Results

About this item

 :
Density:1,200
Storage Temperature:Ambient
UN:1789
ADR:8,II
 :36.46 g/mol
 :GHS05
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324