L(+)-Calcium lactate pentahydrate ≥98.0% USP, Ph. Eur., FCC
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:5743-47-5
Formula:C₆H₁₀CaO₆·5H₂O
MDL Number:MFCD00150344
:308.3 g/mol
Storage Temperature:Ambient
Specification Test Results
- Assay (Dried Basis):98.0 - 101.0%
- Loss On Drying:22.0 - 27.0%
- Volatile Fatty Acids:To Pass Test
- Heavy Metals:max. 10 ppm
- Lead (Pb):max. 2 mg/kg