L-Aspartic acid monosodium salt monohydrate ≥98.0%
Catalog # SPCMA1375-50KGBL
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Fiber drum
- Pk:50 kg
- Seq:4418
Specifications
Specification Test Results
- Assay:98.0 - 101.0%
- Specific Rotation [A]D:+19.0 to +21.0
- Transmittance:95.0 %
- Chloride (Cl):max. 0.04 %
- Ammonium (NH₄):max. 0.04 %
Specification Test Results
About this item
:
:173.11 g/mol (155.10 anhy)
Storage Temperature:Ambient
CAS Number:323194-76-9
Formula:C₄H₆NNaO₄·H₂O
MDL Number:MFCD00152960