Order Entry
Germany
ContactUsLinkComponent
N-Nitroso-N-methylurea
N-Nitroso-N-methylurea
Catalog # SPCMN2939-5GM
Supplier:  Spectrum Chemical
CAS Number:  
N-Nitroso-N-methylurea
Catalog # SPCMN2939-5GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
UN:2811
ADR:6.1,III
 :GHS06
 :GHS07
 :GHS08
Storage Temperature:Refrigerator
CAS Number:684-93-5
Formula:C₂H₅N₃O₂
MDL Number:MFCD00014794