Ammonium iron (III) citrate, brown FCC
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:1185-57-5
:
Formula:C₆H₅+4y)Fe(x)N(y)O₇
MDL Number:MFCD00013099
Boiling Pt:197 °C (1013 hPa)
Density:1,8 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:214-686-6
Specification Test Results
- Assay:16.5% Min.