α-Acetyl-γ-butyrolactone
Catalog # SPCMA2107-200LTBL
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Drum
- Pk:200 L
- Seq:5549
Specifications
About this item
:
:128.13 g/mol
Density:1,190
Storage Temperature:Ambient
CAS Number:517-23-7
Formula:C₆H₈O₃
MDL Number:MFCD00005394