Benzyl nicotinate
Catalog # SPCMB1212-1KGBL
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:213.24 g/mol
Boiling Pt:179…180 °C (8 hPa)
Melting Pt:24 °C
Density:11,165
Storage Temperature:Ambient
EINECS:202-332-3
:GHS07
CAS Number:94-44-0
Formula:C₁₃H₁₁NO₂
MDL Number:MFCD00023584