myo-Inositol ≥97.0% FCC
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:87-89-8
:
Formula:C₆H₁₂O₆
MDL Number:MFCD00077932
:180.16 g/mol
Storage Temperature:Ambient
Specification Test Results
- Assay (Dried Basis):0.97
- Melting Range:224 - 227 °C
- Calcium (Ca):To Pass Test
- Chloride (Cl):max. 0.005 %
- Lead (Pb):max. 4 mg/kg