0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
DL-Sodium lactate
DL-Sodium lactate solution 60% (w/w) in aqueous solution USP
DL-Sodium lactate solution 60% (w/w) in aqueous solution USP
:
Spectrum Chemical
DL-Sodium lactate solution 60% (w/w) in aqueous solution USP
:
Spectrum Chemical
:
72-17-3
:
312-85-6
:
H₃CCH(OH)COONa
:
MFCD00065400
:
112.06 g/mol
:
1,270
:
Ambient
:
200-772-0
Assay (%):
58.8