Dimethyl carbonate ≥99%
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:616-38-6
:
Formula:C₃H₆O₃
MDL Number:MFCD00008420
:90.08 g/mol
Merck Index:12,06117
Boiling Pt:90 °C (1013 hPa)
Melting Pt:0,5…4,7 °C
Density:1,069
Flash Pt:18 °C
Storage Temperature:Ambient
EINECS:210-478-4
UN:1161
ADR:3,II
:GHS02
Specification Test Results
- Assay:99% Min.