Order Entry
Germany
ContactUsLinkComponent
Potassium iodide ≥99.0% USP
Potassium iodide ≥99.0% USP
  SPCMPO185-125GM
 :  
Potassium iodide ≥99.0% USP
  SPCMPO185-125GM
 :  

 

  • Pack type:
    Glass, amber
  • Pk:
    125 g
  • Seq:
    3445

 

 

  • Assay:
    99.0 - 101.5%
  • Alkalinity:
    To Pass Test
  • Loss On Drying:
    max. 1.0 %
  • Iodate:
    max. 4 Ug/G
  • Nitrate, Nitrite, and Ammonia:
    To Pass Test

 

 

 :
 :166 g/mol
 :GHS07
 :GHS08
 :Ambient
 :7681-11-0
 :Kl
 :MFCD00011405