ContactUsLinkComponent
Cole-Parmer® BS-200 Cell Culture Stirring Systems (Stirrers)
Cole-Parmer® BS-200 Cell Culture Stirring Systems (Stirrers)
Supplier:  Cole-Parmer
Certificates
undefined
Cole-Parmer® BS-200 Cell Culture Stirring Systems (Stirrers)
Supplier:  Cole-Parmer
Specifications
  • Catalog No:
  • TECN21837-03
  • TECN21837-04
  • TECN21837-01
  • TECN21837-02
  • No. of stirring positions:
  • 4
  • 4
  • 2
  • 4
  • Max. stirring capacity H₂O:
  • 0,5 L
  • 5 L
  • 1 L
  • 1 L
  • Speed range:
  • 0 - 80 min⁻¹
  • 0 - 80 min⁻¹
  • 0 - 80 min⁻¹
  • 0 - 80 min⁻¹
  • Overall dimensions:
  • 365×250×90 mm
  • 595×530×90 mm
  • 400×140×90 mm
  • 495×395×90 mm
  • Timer:
  • Variable on time: 6 seconds to 5 minutes; Variable off time: 2 minutes to 2 hours
  • Variable on time: 6 seconds to 5 minutes; Variable off time: 2 minutes to 2 hours
  • Variable on time: 6 seconds to 5 minutes; Variable off time: 2 minutes to 2 hours
  • Variable on time: 6 seconds to 5 minutes; Variable off time: 2 minutes to 2 hours
  • Electrical:
  • 120/230 VAC, 50/60 Hz
  • 120/230 VAC, 50/60 Hz
  • 120/230 VAC, 50/60 Hz
  • 120/230 VAC, 50/60 Hz
Specifications
Related Information
Culture vessels
Product Information