Specifications
- Antibody type:Primary
- Antigen name:C1orf69
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:131-230/256
- Form:Liquid
- Gene ID:200205
- Antigen synonyms:Iron-sulfur cluster assembly factor homolog|FLJ12734|IBA57 iron sulfur cluster assembly homolog|C1orf69|Putative transferase C1orf69 mitochondrial|FLJ13849|Mitochondrial proteins with a mass of 57kDa|Putative transferase C1orf69|Iron sulfur cluster assembly factor for biotin synthase and aconitase like|mitochondrial|Putative transferase CAF17, mitochondrial|CA069_HUMAN|IBA57 iron sulfur cluster assembly homolog S. cerevisiae|CAF17_HUMAN|IBA57
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:34 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C1orf69
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Belonging to the CAF17 subfamily of the gcvT family, C1orf69 is a 356 amino acid mitochondrial protein that is required for normal heme synthesis. Heme synthesis is dependent upon receiving iron through iron sulfur (Fe-S) cluster biogenesis proteins such as C1orf69, which is specifically thought to assemble Fe-S clusters for mitochondrial aconitase and lipoate synthase. In zebrafish, deficiency of C1orf69 causes anemia. The gene encoding C1orf69 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1.
Type: Primary
Antigen: C1orf69
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 131-230/256
Host: Rabbit
Isotype: IgG
Reactivity: Rat