Order Entry
Germany
ContactUsLinkComponent
Anti-Golgi phosphoprotein 6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-Golgi phosphoprotein 6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-13492R-A750
Supplier:  Bioss
Anti-Golgi phosphoprotein 6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-13492R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Golgi phosphoprotein 6
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Xenopus laevis,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    151-250/452
  • Form:
    Liquid
  • Gene ID:
    26003
  • Antigen synonyms:
    GORASP2|Golgi phosphoprotein 6|GOLPH2|GOLPH6|GORS2_HUMAN|GRASP55|Golgi reassembly-stacking protein of 55 kDa|GRS2|Golgi reassembly stacking protein of 55 kDa|Golgi reassembly stacking protein 2|Golgi reassembly-stacking protein 2|p59|Golgi reassembly stacking protein 2, 55kDa|RP23-428O15.1
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    47 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Golgi phosphoprotein 6
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

The Golgi apparatus is a highly complex organelle comprised of a stack of cisternal membranes on the secretory pathway from the ER to the cell surface. The structure is maintained by an exoskeleton or Golgi matrix constructed from a family of coiled-coil protein, the golgins and other peripheral membrane components such as GRASP55 and GRASP65 (1). GRASP55 (Golgi reassembly stacking protien or p59) is a component of the Golgi stacking machinery. GRASP55 is highly homologous to GRASP65 and contains two PDZ domains. GRASP55 is myristoylated and palmitoylated. Unlike GRASP65, GRASP55 does not have detectable binding with the vesicle docking protein GM130 and is located on the medial-Golgi rather than cis-Golgi. Both GRASP55 and GRASP65 function in the stacking of Golgi Cisternae (2,3). The novel coiled-coil protein golgin 45 interacts with GRASP55 and the GTP form of Rab 2, suggesting that GRASP55 and golgin 45 form a Rab 2 effector complex on medial-Golgi essential for normal protein transport and Golgi structure (4). ERK2 directly phosphorylates GRASP55, which is phosphorylated in mitotic cells, suggesting that mitogen-activated protein kinase kinase (MKK)/ERK pathway phosphorylates the Golgi during mitosis (5).

Type: Primary
Antigen: Golgi phosphoprotein 6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 151-250/452
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Xenopus laevis