Order Entry
Germany
ContactUsLinkComponent
Prostaglandin Screening Library II
Prostaglandin Screening Library II
Catalog # CAYM10502-50
Supplier:  Cayman Chemical
CAS Number:  
undefined
Prostaglandin Screening Library II
Catalog # CAYM10502-50
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    50 µl
  • Description:
    Prostaglandins in a 96-well matrix tube rack format

Specifications

About this item

For D and E series prostaglandin analog screening.

  • Contains >60 compounds
  • Includes the ''D- and E-series'' prostaglandins

The Prostaglandin Screening Library II contains more than 60 ''D and E-series'' of prostaglandins in a 96-well Matrix tube rack format as 2 mM stocks in DMSO. The composition of this screening library will always vary somewhat depending upon our inventory. Stability data is not available for compounds as supplied in the screening library.