Anti-GDPD5 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-9207R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:GDPD5
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:301-400/605
- Form:Liquid
- Gene ID:81544
- Antigen synonyms:GDE2|Gdpd5|GDPD5_HUMAN|Glycerophosphodiester phosphodiesterase domain-containing protein 5|Glycerophosphodiester phosphodiesterase domain containing 5|Glycerophosphodiester phosphodiesterase 2|PP1665
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GDPD5
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Promotes neurite formation. Cooperates with PRDX1 to drive postmitotic motor neuron differentiation. The glycerophosphodiester phosphodiesterase activity may be required for its role in neuronal differentiation. May contribute to the osmotic regulation of cellular glycerophosphocholine.
Type: Primary
Antigen: GDPD5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/605
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat