Order Entry
Germany
ContactUsLinkComponent
Anti-Eph receptor A2+A3+A4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-Eph receptor A2+A3+A4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-7553R-A750
Supplier:  Bioss
Anti-Eph receptor A2+A3+A4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-7553R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Eph receptor A2+A3+A4
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    751-850/976
  • Form:
    Liquid
  • Gene ID:
    1969
  • Antigen synonyms:
    TYRO4|EPHA2|EPHA3|EPHA3_HUMAN|HEK|EPH Receptor A3|ETK1|HEK4|HEK8|EPHA4|EPH Receptor A4|SEK|ECK|EPH Receptor A2|EPHA2_HUMAN|EPHA4_HUMAN
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    105 - 125 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Eph receptor A2+A3+A4
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Eph receptor A2 is the receptor for members of the ephrin-A family. It binds to ephrin-A1, -A3, -A4 and -A5. Eph receptor A3 is also a receptor for members of the ephrin-A family. It binds to ephrin-A2, -A3, -A4 and -A5 and is thought to play a role in lymphoid function. Eph receptor A4 is a receptor for members of the ephrin-A family. It binds to ephrin-A1, -A4 and -A5. It binds more poorly to ephrin-A2 and -A3. It may play a role in a signal transduction process involved in hindbrain pattern formation.

Type: Primary
Antigen: Eph receptor A2+A3+A4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 751-850/976
Host: Rabbit
Isotype: IgG
Reactivity: