Order Entry
Germany
ContactUsLinkComponent
Anti-Adenylosuccinate Lyase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-Adenylosuccinate Lyase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-6352R-A750
Supplier:  Bioss
Anti-Adenylosuccinate Lyase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-6352R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Adenylosuccinate Lyase
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Epitope:
    185-280/484
  • Form:
    Liquid
  • Antigen synonyms:
    Adenylosuccinase|ASL|Adenylosuccinate lyase|PUR8_HUMAN|AMPS|ASase|ADSL
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    55 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Adenylosuccinate Lyase
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Adenylsuccinate lyase is involved in both de novo synthesis of purines and formation of adenosine monophosphate from inosine monophosphate. It catalyzes two reactions in AMP biosynthesis: the removal of a fumarate from succinylaminoimidazole carboxamide (SAICA) ribotide to give aminoimidazole carboxamide ribotide (AICA) and removal of fumarate from adenylosuccinate to give AMP. Adenylosuccinase deficiency results in succinylpurinemic autism, psychomotor retardation, and , in some cases, growth retardation associated with muscle wasting and epilepsy. Two transcript variants encoding different isoforms have been found for this gene.

Type: Primary
Antigen: Adenylosuccinate Lyase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 185-280/484
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse