Order Entry
Germany
ContactUsLinkComponent
Anti-SNF5 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-SNF5 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6109R-A680
Supplier:  Bioss
Anti-SNF5 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6109R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SNF5
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Epitope:
    301-385/385
  • Form:
    Liquid
  • Gene ID:
    6598
  • Antigen synonyms:
    SNF5|hSNF5|SWI/SNF complex component SNF5|TYE4|Transcription regulatory protein SNF5|SNF5L1|Snr1|SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily B member 1|RDT|Transcription factor TYE4|INI1 antibody Integrase interactor 1 protein|Sfh1p|SWI10|BAF47|SMARCB1|Sucrose nonfermenting yeast homolog like 1
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    44 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human SNF5
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

SNF5 is involved in transcriptional activation. The SWI/SNF complex is required for the induced expression of a large number of genes. This complex alters chromatin structure to facilitate binding of gene-specific dedicated transcription factors. SNF5 also funtions as a tumor suppressor gene and is frequently mutated in malignant rhabdoid tumors. It is also involved in adipocyte differentiation.

Type: Primary
Antigen: SNF5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-385/385
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse