Anti-MTUS1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6076R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:MTUS1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:1151-1270/1270
- Form:Liquid
- Gene ID:57509
- Antigen synonyms:GK1|Mitochondrial tumor suppressor 1|ATBP|ATIP|Microtubule-associated tumor suppressor 1|MP44|ICIS|Angiotensin-II type 2 receptor-interacting protein|KIAA1288|AT2 receptor-binding protein|MTUS1|MTSG1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:141 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human MTUS1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Cooperates with AGTR2 to inhibit ERK2 activation and cell proliferation. May be required for AGTR2 cell surface expression. Together with PTPN6, induces UBE2V2 expression upon angiotensin-II stimulation. Isoform 1 inhibits breast cancer cell proliferation, delays the progression of mitosis by prolonging metaphase and reduces tumor growth.
Type: Primary
Antigen: MTUS1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1151-1270/1270
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat