Anti-Bcl2L2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-5903R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Bcl2L2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:1-100/193
- Form:Liquid
- Gene ID:599
- Antigen synonyms:Bcl2 L2|BCL2 like 2 protein|BCL 2 Like 2|Bcl2-L-2|BCL2 like 2|Bcl2l2|Bcl-2-like protein 2|Bcl 2L2|Bcl 2 like 2 protein|Apoptosis regulator Bcl-W|BCL W|BCLW|Apoptosis regulator BCL W|B2CL2_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:21 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Bcl2L2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Promotes cell survival. Blocks dexamethasone-induced apoptosis. Mediates survival of postmitotic Sertoli cells by suppressing death-promoting activity of BAX.
Type: Primary
Antigen: Bcl2L2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/193
Host: Rabbit
Isotype: IgG
Reactivity: Mouse