Safety Waste Collectors
Supplier: S.C.A.T.
Certificates
About this item
The Safety Waste Collector connects capillaries and tubes of the HPLC system with the disposal system - suitable for the SymLine benchtop feedthroughs and SymLine FLEX.
- For collecting capillaries and tubes in a cap on the disposal container.
- Various number of connections on one cap with standardised GL45 threads
- Available in electrically conductive (black core) and non-electrically conductive (white core) version
SymLine is the modular disposal system for liquid waste (e.g. solvents, acids, alkalis, etc.). It connects several laboratory workstations, HPLC systems and fume cupboards to a central disposal or collection point, such as a safety cabinet.