Order Entry
Germany
ContactUsLinkComponent
Anti-AMPD3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-AMPD3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-9517R-A750
Supplier:  Bioss
Anti-AMPD3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-9517R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AMPD3
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    21-120/767
  • Form:
    Liquid
  • Gene ID:
    272
  • Antigen synonyms:
    Erythrocyte type AMP deaminase|AMP deaminase 3|Erythrocyte AMP deaminase|Ampd3|Myoadenylate deaminase|Adenosine monophosphate deaminase isoform E|AMPD3_HUMAN|Erythrocyte specic AMP deaminase|AMP aminohydrolase|Adenosine monophosphate deaminase 3|AMP deaminase isoform E
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human AMPD3
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

AMP deaminase plays a critical role in energy metabolism. Involvement in diseaseDefects in AMPD3 are the cause of adenosine monophosphate deaminase deficiency erythrocyte type (AMPDDE); also known as erythrocyte AMP deaminase deficiency. AMPDDE is a metabolic disorder due to lack of activity of the erythrocyte isoform of AMP deaminase. It is a clinically asymptomatic condition characterised by a 50% increase in steady-state levels of ATP in affected cells. Individuals with complete deficiency of erythrocyte AMP deaminase are healthy and have no hematologic disorders.

Type: Primary
Antigen: AMPD3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/767
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat