Anti-RGS4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-9452R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:RGS4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:51-150/205
- Form:Liquid
- Gene ID:5999
- Antigen synonyms:Schizophrenia disorder 9|MGC60244|RGP 4|MGC2124|Regulator of G-protein signaling 4|RGS4_HUMAN|SCZD9|RGS4|RGS 4|Regulator of G protein signalling 4|RGP-4|SCZD 9
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human RGS4
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. Activity on G(z)-alpha is inhibited by phosphorylation of the G-protein. Activity on G(z)-alpha and G(i)-alpha-1 is inhibited by palmitoylation of the G-protein.
Type: Primary
Antigen: RGS4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/205
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat