Anti-Legumain Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # BOSSBS-3907R-PE
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Legumain
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:201-300/433
- Form:Liquid
- Gene ID:5641
- Antigen synonyms:AEP|Protease, cysteine 1|Legumain|Asparaginyl endopeptidase|LGMN1|LGMN|PRSC1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:46 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Legumain
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Has a strict specificity for hydrolysis of asparaginyl bonds. Can also cleave aspartyl bonds slowly, especially under acidic conditions. Required for normal lysosomal protein degradation in renal proximal tubules. Required for normal degradation of internalized EGFR. Plays a role in the regulation of cell proliferation via its role in EGFR degradation (By similarity). May be involved in the processing of proteins for MHC class II antigen presentation in the lysosomal/endosomal system.
Type: Primary
Antigen: Legumain
Clonality: Polyclonal
Clone:
Conjugation: PE
Public Immunogen Range: 201-300/433
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat