Anti-dUTPase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-1721R-A750
: Bioss
- Antibody type:Primary
- Antigen name:dUTPase
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:151-252/252
- Form:Liquid
- Gene ID:1854
- Antigen synonyms:DUT|dUTP pyrophosphatase|dUTPase|Deoxyuridine 5'-triphosphate nucleotidohydrolase, mitochondrial
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:20 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human dUTPase
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
This enzyme is involved in nucleotide metabolism: it produces dUMP, the immediate precursor of thymidine nucleotides and it decreases the intracellular concentration of dUTP so that uracil cannot be incorporated into DNA.
Type: Primary
Antigen: dUTPase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 151-252/252
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse