Anti-B3GAT1/CD57 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-1635R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:B3GAT1/CD57
- Clonality:Polyclonal
- Conjugation:Cy7®
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:21-120/334
- Form:Liquid
- Gene ID:27087
- Antigen synonyms:LEU7|GLCATP|CD57|NK1|GLCUATP|NK-1|GlcUAT-P|GlcAT-P|Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1|Beta-1,3-glucuronyltransferase 1|Glucuronosyltransferase P|UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase|B3GAT1|HNK1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:38 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human B3GAT1/CD57
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Involved in the biosynthesis of L2/HNK-1 carbohydrate epitope on glycoproteins. Can also play a role in glycosaminoglycan biosynthesis. Substrates include asialo-orosomucoid (ASOR), asialo-fetuin, and asialo-neural cell adhesion molecule. Requires sphingomyelin for activity: stearoyl-sphingomyelin was the most effective, followed by palmitoyl-sphingomyelin and lignoceroyl-sphingomyelin. Activity was demonstrated only for sphingomyelin with a saturated fatty acid and not for that with an unsaturated fatty acid, regardless of the length of the acyl group (By similarity).
Type: Primary
Antigen: B3GAT1/CD57
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 21-120/334
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat