Anti-RGS2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-0492R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:RGS2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Epitope:116-211/211
- Form:Liquid
- Gene ID:5997
- Antigen synonyms:GS8|G/G1 switch regulatory protein 8|RGS2|Cell growth-inhibiting gene 31 protein|GIG31|Regulator of G-protein signaling 2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:24 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human RGS2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. May play a role in leukemogenesis. Plays a role in negative feedback control pathway for adenylyl cyclase signaling. Binds EIF2B5 and blocks its activity, thereby inhibiting the translation of mRNA into protein.
Type: Primary
Antigen: RGS2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 116-211/211
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat