Order Entry
Germany
ContactUsLinkComponent
Anti-HOXA5 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-HOXA5 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-5713R-A680
Supplier:  Bioss
Anti-HOXA5 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-5713R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HOXA5
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    201-270/270
  • Form:
    Liquid
  • Gene ID:
    3202
  • Antigen synonyms:
    Homeo box 1C antibody Homeobox A5 antibody Homeobox protein HOXA5 antibody HOX 1 antibody HOX 1.3 antibody HOX 1C antibody HOX1 antibody HOX1.3 antibody HOX1C antibody|HXA5_HUMAN
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    29 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human HOXA5
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

HOXA5 upregulates both p53 promoter-reporter constructs and endogenous p53 synthesis, leading to apoptosis. HOXA5 is detectable in approximately one-third of primary tumors. Lack of HOXA5 expression strongly correlates with methylation of its promoter region, suggesting a causal role for methylation in the silencing of HOXA5 gene expression. HOXA5 expression is an important step in tumorigenesis and loss of expression of p53 in human breast cancer may be primarily due to lack of expression of HOXA5.

Type: Primary
Antigen: HOXA5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-270/270
Host: Rabbit
Isotype: IgG
Reactivity: