Anti-YES1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-5592R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:YES1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:10413
- Antigen synonyms:p61-Yes|YES1phospho Y537|Proto-oncogene tyrosine-protein kinase Yes|Proto oncogene tyrosine protein kinase YES|c Yes|HsT441|Yamaguchi sarcoma oncogene|Yes 1|v yes 1 Yamaguchi sarcoma viral oncogene homolog 1|YES1|Yes|Proto-oncogene c-Yes|Viral oncogene yes1 homolog 1|Cellular Yes1 protein|c-Yes|Viral oncogene yes 1 homolog 1|YES_HUMAN|P61 YES|Cellular Yes 1 protein
- Modification:Phosphorylation (Tyr537)
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:60 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated Synthesised phosphopeptide derived from human YES1 around the phosphorylation site of Tyr537:PQ(p-Y)QP?
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This gene is the cellular homolog of the Yamaguchi sarcoma virus oncogene. The encoded protein has tyrosine kinase activity and belongs to the src family of proteins. This gene lies in close proximity to thymidylate synthase gene on chromosome 18, and a corresponding pseudogene has been found on chromosome 22.
Type: Primary
Antigen: YES1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: