Order Entry
Germany
ContactUsLinkComponent
Anti-CD73 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-CD73 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-4834R-A750
Supplier:  Bioss
Anti-CD73 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-4834R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD73
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Sheep,
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    151-250/574
  • Form:
    Liquid
  • Gene ID:
    4907
  • Antigen synonyms:
    Ecto-5'-nucleotidase|5'-NT|eN|E5NT|NTE|NT5|NT5E|5'-nucleotidase|CALJA|eNT|NT|CD73
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    63 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human CD73/NT5
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Hydrolyses extracellular nucleotides into membrane permeable nucleosides. Exhibits AMP-, NAD-, and NMN-nucleosidase activities.

Type: Primary
Antigen: CD73
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 151-250/574
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat; Bovine; Dog; Sheep