Anti-Methionine adenosyltransferase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-4054R-A680
: Bioss
- Antibody type:Primary
- Antigen name:Methionine adenosyltransferase
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:31-130/395
- Form:Liquid
- Gene ID:4143
- Antigen synonyms:MAT 2A|SAMS2|Methionine adenosyltransferase II|Methionine adenosyltransferase 2|S adenosylmethionine synthetase gamma form|METK1_HUMAN|S adenosylmethionine synthetase isoform type 2|AMS2|Methionine adenosyltransferase II alpha|AdoMet synthetase|MATA 2|MAT II|MATA2|MAT2A AdoMet synthetase 2|MATII|AMS 2|Methionine adenosyltransferase|SAMS 2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:44 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Methionine adenosyltransferase
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
S adenosylmethionine synthetase catalyses the formation of S adenosylmethionine from methionine and ATP. In mammalian tissues, there are three distinct forms of AdoMet synthases designated as alpha, beta, and gamma. Alpha and beta are expressed only in adult liver, while gamma is widely distributed in extrahepatic tissues.
Type: Primary
Antigen: Methionine adenosyltransferase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 31-130/395
Host: Rabbit
Isotype: IgG
Reactivity: Rat