Anti-Pancreatic Amylase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-4030R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Pancreatic Amylase
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:151-250/511
- Form:Liquid
- Gene ID:279
- Antigen synonyms:PA|AMY2|Pancreatic alpha-amylase|AMY2A|1,4-alpha-D-glucan glucanohydrolase
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:56 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Pancreatic Amylase
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Pancreatic amylase is a digestive enzyme secreted by the pancreas. It consists of a single polypeptide chain with a molecular weight of 54 kDa.Pancreatic amylase and salivary amylase are the products of separate genes and, although they share 98% amino acid sequence homology, they differ in molecular size, isoelectric point and antigenic properties. Measurement of amylase concentrations in serum is a diagnostic test for various pancreatic disorders (especially acute and relapsing pancreatitis).
Type: Primary
Antigen: Pancreatic Amylase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 151-250/511
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat