Order Entry
Germany
ContactUsLinkComponent
Anti-RGS5 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-RGS5 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2794R-A680
Supplier:  Bioss
Anti-RGS5 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2794R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RGS5
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Epitope:
    81-181/181
  • Form:
    Liquid
  • Gene ID:
    19737
  • Antigen synonyms:
    RGS5|Regulator of G-protein signaling 5|1110070A02Rik
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    22 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from mouse RGS5
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. Binds to G(i)-alpha and G(o)-alpha, but not to G(s)-alpha.

Type: Primary
Antigen: RGS5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 81-181/181
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat