Specifications
- Antibody type:Primary
- Antigen name:MuRF1/Trim63
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:251-353/353
- Form:Liquid
- Gene ID:84676
- Antigen synonyms:MuRF 1|Iris ring finger protein|Muscle specic ring finger protein 2|Tripartite mot containing protein 63|MURF1|IRF|TRIM 63|FLJ32380|MuRF-1|SMRZ|Ubiquitin ligase TRIM63|RNF28|TRIM63|MURF2|E3 ubiquitin-protein ligase TRIM63|Striated muscle RING zinc finger protein|MURF 1|Muscle-specic RING finger protein 1|Ring finger protein 28|Tripartite mot containing 63
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:39 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human MuRF1/Trim63
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This gene encodes a member of the RING zinc finger protein family found in striated muscle and iris. The product of this gene is localised to the Z-line and M-line lattices of myofibrils, where titin's N-terminal and C-terminal regions respectively bind to the sarcomere. In vitro binding studies have shown that this protein also binds directly to titin near the region of titin containing kinase activity. Another member of this protein family binds to microtubules. Since these family members can form heterodimers, this suggests that these proteins may serve as a link between titin kinase and microtubule-dependent signal pathways in muscle.
Type: Primary
Antigen: MuRF1/Trim63
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 251-353/353
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat