Anti-BPNT1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Catalog # BOSSBS-20136R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:BPNT1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:2-100/308
- Form:Liquid
- Gene ID:10380
- Antigen synonyms:3'(2'),5'-bisphosphate nucleotidase 1|PIP|BPNT1|Bisphosphate 3'-nucleotidase 1|PAP-inositol 1,4-phosphatase|HEL20
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:33 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human BPNT1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Converts adenosine 3'-phosphate 5'-phosphosulfate (PAPS) to adenosine 5'-phosphosulfate (APS) and 3'(2')-phosphoadenosine 5'- phosphate (PAP) to AMP. Has 1000-fold lower activity towards inositol 1,4-bisphosphate (Ins(1,4)P2) and inositol 1,3,4-trisphosphate (Ins(1,3,4)P3), but does not hydrolyse Ins(1)P, Ins(3,4)P2, Ins(1,3,4,5)P4 or InsP6.
Type: Primary
Antigen: BPNT1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 2-100/308
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat