Anti-CCL26 Rabbit Polyclonal Antibody (FITC)
BOSSBS-15513R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:CCL26
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:21-94/94
- Form:Liquid
- Gene ID:685958
- Antigen synonyms:Small inducible cytokine subfamily A Cys member 26|TSC1|C-C mot chemokine 26|MIP 4alpha|Small inducible cytokine A26|Chemokine C C mot ligand 26|MIP 4a|SCYA 26|Macrophage inflammatory protein 4-alpha|IMAC|Eotaxin-3|SCYA26|Thymic stroma chemokine 1|Small inducible cytokine subfamily A member 26|CCL26
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:8 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from rat CCL26
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Eotaxin 3 is a recently discovered CC chemokine. It binds to the CCR3 receptor and is highly chemotactic for eosinophils and basophils.
Type: Primary
Antigen: CCL26
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 21-94/94
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat