Anti-CCL26 Rabbit Polyclonal Antibody (FITC)
BOSSBS-15512R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:CCL26
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:51-94/94
- Form:Liquid
- Antigen synonyms:TSC1|Small inducible cytokine subfamily A Cys Cys member 26|C-C mot chemokine 26|TSC-1|MIP 4alpha|MIP4alpha|Small inducible cytokine A26|Chemokine C C mot ligand 26|MIP 4a|TSC 1|SCYA 26|Thymic stroma chemokine-1|Macrophage inflammatory protein 4-alpha|MIP-4-alpha|CCL 26|CCL26_HUMAN|IMAC|CC chemokine IMAC|Eotaxin-3|SCYA26|Thymic stroma chemokine 1|Small-inducible cytokine A26|Small inducible cytokine subfamily A member 26|MIP4a|CCL26
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:8 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CCL26
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
CCL26 is a recently discovered CC chemokine. It binds to the CCR3 receptor and is highly chemotactic for eosinophils and basophils.
Type: Primary
Antigen: CCL26
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 51-94/94
Host: Rabbit
Isotype: IgG
Reactivity: Human