Anti-COMT Rabbit Polyclonal Antibody (HRP)
Catalog # BOSSBS-15507R-HRP
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:COMT
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Epitope:121-220/271
- Form:Liquid
- Gene ID:1312
- Antigen synonyms:COMT|Catechol O-methyltransferase|HEL-S-98n
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:30 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human COMT
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Catalyses the O-methylation, and thereby the inactivation, of catecholamine neurotransmitters and catechol hormones. Also shortens the biological half-lives of certain neuroactive drugs, like L-DOPA, alpha-methyl DOPA and isoproterenol.
Type: Primary
Antigen: COMT
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 121-220/271
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat