Anti-TEC Rabbit Polyclonal Antibody (FITC)
Catalog # BOSSBS-15505R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:TEC
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:11-100/631
- Form:Liquid
- Gene ID:7006
- Antigen synonyms:PSCTK4|Tec protein tyrosine kinase|Tec|Tyrosine protein kinase|MGC126762|MGC126760|TEC_HUMAN|Tyrosine-protein kinase Tec
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:74 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TEC/Tec protein tyrosine kinase
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
TEC is a belongs to the Tec family of non-receptor protein-tyrosine kinases containing a pleckstrin homology domain. Tec family kinases are involved in the intracellular signaling mechanisms of cytokine receptors, lymphocyte surface antigens, heterotrimeric G-protein coupled receptors, and integrin molecules. They are also key players in the regulation of the immune functions. Tec kinase is an integral component of T cell signaling and has a distinct role in T cell activation. This gene may be associated with myelodysplastic syndrome.
Type: Primary
Antigen: TEC
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 11-100/631
Host: Rabbit
Isotype: IgG
Reactivity: