Anti-EXOC2/SEC5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-13118R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:EXOC2/SEC5
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:31-130/924
- Form:Liquid
- Gene ID:55770
- Antigen synonyms:Sec5p|Exocyst complex component 2|Exocyst complex component Sec5|SEC5L1|SEC5|EXOC2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:104 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human EXOC2/SEC5
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Component of the exocyst complex involved in the docking of exocytic vesicles with fusion sites on the plasma membrane.
Type: Primary
Antigen: EXOC2/SEC5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 31-130/924
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat