Anti-COL3A1 Rabbit Polyclonal Antibody
- Catalog No:
- ROCK600-401-105-05
- ROCK600-401-105-01
- Antigen symbol:
- COL3A1
- COL3A1
- Antigen name:
- Collagen alpha-1(III) chain
- Collagen alpha-1(III) chain
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Pig, Human, Bovine
- Pig, Human, Bovine
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 1281
- 1281
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Collagen Type III from human and bovine placenta
- Collagen Type III from human and bovine placenta
- Format:
- IgG
- IgG
- Form:
- Liquid (sterile filtered)
- Liquid (sterile filtered)
- Concentration:
- 1.16 mg/mL by UV absorbance at 280 nm
- 1.16 mg/mL by UV absorbance at 280 nm
- Storage buffer:
- 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- Storage temperature:
- 4°C
- 4°C
- Shipping temperature:
- 20
- 20
- Tested applications:
- Dot Blot
- Dot Blot