ESD Safety Shoes, Strap Closure, Low, S3
: ELTEN
ESD safety shoes feature a Velcro® closure. Available in white finish.
- Hydrophobised microfibre
- Fabric lining for natural ventilation
- Full, padded bellows tongue
- Full-length inlay sole ESD PRO grey
- MONO-PU sole SAFETY-GRIP
- Steel toe cap
Safety shoes feature a non-metallic, penetration-resistant midsole.
: EN ISO 20345 S3 SRC, form A.
- Catalog No:
- ELTN720171/43
- ELTN720171/45
- ELTN720171/44
- ELTN720171/47
- ELTN720171/46
- ELTN720171/48
- ELTN720171/40
- ELTN720171/39
- ELTN720171/42
- ELTN720171/41
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001