Order Entry
Germany
ContactUsLinkComponent
QuickDetect™ Free Cholesterol (fCHOL) (Mouse) ELISA Kit
QuickDetect™ Free Cholesterol (fCHOL) (Mouse) ELISA Kit
Catalog # ABCAAB285294-96
Supplier:  Abcam
CAS Number:  
undefined
QuickDetect™ Free Cholesterol (fCHOL) (Mouse) ELISA Kit
Catalog # ABCAAB285294-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    QuickDetect™ free cholesterol (fCHOL) (Mouse) ELISA kit, Size; 100 assays

Specifications

About this item

A rapid sandwich ELISA kit to quantify fCHOL in mouse serum, plasma, cell culture supernatnat.

  • Easy, convenient and time-saving method to measure the level of free Cholesterol in mouse serum, plasma, culture supernatants and other biological fluids
  • The entire assay can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results
  • Detection Range: 1,25 to 80 ng/ml

Cholesterol, is an organic molecule, a sterol (or modified steroid), a type of lipid molecule, and is biosynthesised by all animal cells, in which it is an essential structural component of animal cell membranes that is required to maintain both membrane structural integrity and fluidity. Cholesterol enables animal cells to dispense with a cell wall, thereby allowing animal cells to change shape and animals to move. BioVision’s QuickDetect™ fCHOL ELISA kit is a sandwich ELISA assay for measuring the quantity of mouse fCHOL in serum, plasma and cell culture and other biological fluid. The entire process can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.